Learn More
Hampton Research STOCKOPT GLYCINE #04 PH 8.9
SDP
Supplier:  Hampton Research HR225004
Encompass
HR2-250-04/StockOptions Glycine #04, 1.0 M Glycine pH 8.9, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-150