Learn More
Hampton Research STOCKOPT GLYCINE #08 PH 9.3
SDP
Supplier:  Hampton Research HR225008
Encompass
HR2-250-08/StockOptions Glycine #08, 1.0 M Glycine pH 9.3, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-154