Learn More
Hampton Research STOCKOPT MALIC ACID #24 PH 6.0
SDP
Supplier:  Hampton Research HR224024
Encompass
HR2-240-24/StockOptions Malic Acid #24, 1.0 M DL-Malic acid pH 6.0, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-121