Learn More
Hampton Research STOCKOPT MOPS #01 PH 6.5
SDP
Supplier:  Hampton Research HR225201
Encompass
HR2-252-01/StockOptions MOPS #01, 1.0 M MOPS pH 6.5, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-186