Learn More
Hampton Research STOCKOPT MOPS #05 PH 6.9
SDP
Supplier:  Hampton Research HR225205
Encompass
HR2-252-05/StockOptions MOPS #05, 1.0 M MOPS pH 6.9, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-190