Learn More
Hampton Research STOCKOPT TRICINE #08 PH 8.1
SDP
Supplier:  Hampton Research HR225308
Encompass
HR2-253-08/StockOptions Tricine #08, 1.0 M Tricine pH 8.1, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-209