Learn More
Cayman Chemical STOML3 InhIbItr OB-1 5mg
SDP
Supplier:  Cayman Chemical 375845 MG
Encompass_Preferred
An inhibitor of STOML3 oligomerization; inhibits oligomerization of mouse and human STOML3 in cell-free assays at 10 µM; reduces Piezo1 channel current amplitude in Neuro2a cells; inhibits tactile perception in wild-type mice and reduces tactile allodynia in a mouse model of streptozotocin-induced diabetic neuropathy
Catalog No. 50-291-6394