Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarstedt Inc STOP F/TUBE 12MM PINK/PK1000

Supplier:  Sarstedt Inc 65.809.001

Encompass_Preferred

Stopper for Tube 12mm Pink ..

Catalog No. NC9670045


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.