Learn More
BURKLE INC stop-it hose clamp, Easy-Click, Ø 0.39 in. (10 mm), red
Supplier:  BURKLE INC 86190102
Encompass_Preferred
Stepless flow regulation: With the tubing clamp stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections. The tubing is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged.
Catalog No. 50-000-25811