Learn More
BURKLE INC StopCock angled, cap nut 3/4"- spout with nozzle
Supplier:  BURKLE INC 05251000
Encompass_Preferred
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.
The rotatable spout angle prevents dripping. The reduced spout allows the water to flow into narrow openings.
Catalog No. NC2187653