Learn More
HARVARD BIOSCIENCE INC STOPCOCK GREASE DOW CORNING
Supplier:  HARVARD BIOSCIENCE INC 640275
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 12.This convenient package includes a tube of Dow Corning 111 Valve Lubricant & Sealant, two acrylic paint brushes (sized #2 and #4), and several pallets. By "painting" the grease onto the bottom surface of a polycarbonate chamber, it is easy to evenly spread lubricant and create a water-tight seal.
Catalog No. NC0852304