Learn More
Chemglass Life Sciences Stopcock, T-Type, Hollow Glass Plug, High Vacuum, 4mm Bore, 17.35/50 Plug Size, 10mm OD Sidearms
SDP
Supplier:  Chemglass Life Sciences CG48002
Encompass_Preferred
Similar to CG-478, but with the addition of an extra horizontal sidearm. Precision ground and hand-lapped with corresponding numbers on barrel and plug.
Catalog No. 50-873-737