Learn More
Wilmad Labglass STOPPER ASSEM 500ML 29/42 EC
Supplier:  Wilmad Labglass LG3762124
Encompass_Preferred
Bottle, Gas Washing, Tall Form, Fritted. Stopper Only. Capacity: 500mL, Joint Size: [ST] 29/42; Porosity: Extra Coarse. Manufactured with 12mm OD x 25mm length fritted cylinder. Supplied with LG-1004-110 plastic joint clamp.
Catalog No. 50-807-081
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.