Learn More
Wilmad Labglass STOPPER PENNYHEAD SLD ST 19
Supplier:  Wilmad Labglass LG10320106
Encompass_Preferred
Stopper, Penny Head, Solid, Flask Size. Joint Size: [ST] 19. Stopper is used extensively with volumetric flasks and separatory funnels.
Catalog No. 50-802-845