Learn More
Wilmad Labglass STOPPER PENNYHEAD SLD ST 27
Supplier:  Wilmad Labglass LG10320110
Encompass_Preferred
Stopper, Penny Head, Solid, Flask Size. Joint Size: [ST] 27. Stopper is used extensively with volumetric flasks and separatory funnels.
Catalog No. 50-806-352
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.