Learn More
Chemglass Life Sciences 29/42 Stopper, Hollow
SDP
Supplier:  Chemglass Life Sciences CG3002L29
Encompass_Preferred
29/42 Stopper, Hollow. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.
Catalog No. CG3002L29