Learn More
Grainger Stopper Sz 2 16 mm BDia Black 0 PK25
Supplier:  Grainger 16WC93
Encompass_Preferred
The Grainger Approved 2-1H Stopper Sz 2, 16 mm BDia, Black 0 PK25 is a rubber stopper made of black butyl rubber. It has an outer diameter of 16 mm, an inner diameter of 6 mm, and a length of 16 mm. This stopper is designed to provide a tight seal and fit snugly into containers. It comes in a pack of 25 stoppers. The stoppers are rated to withstand temperatures between -65 to 250°F. This product can be used to plug bottles, vials, tubes and other containers in laboratory, industrial, and chemical processing applications. The black color provides contrast for easy visual inspection.
Catalog No. 50-276-4850