Learn More
CELLTREAT Scientific Products Storage Box, 50mL Centrifuge Tubes, 16 Place, Polypropylene, Case Quantity 4
SDP
Supplier:  CELLTREAT Scientific Products 229947
Encompass_Preferred
Conveniently store 15 and 50mL Centrifuge Tubesin temperatures as low as -80°C
Durable polypropylene Storage Boxes are fullyautoclavable
Integrated rack features printed numericalcoordinates
Safely and easily remove boxes from freezer racksusing textured pull tab
Catalog No. 50-006-70054