Learn More
Research Products International Corp Storage Rack for Upright Freezers with Locking Rods, Holds 96-Well or 384-Well Plates, Rack Dimensions (Inches) 5 1/2 x 18 1/2 x 11H
SDP
Supplier:  Research Products International Corp 247551LR
Encompass
Fits micro-plates with dimensions of 5" x 3-3/8" x 9/16" H
Catalog No. 50-197-7646