Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, PTFE Plug, 6mm Bore, 20/65 Plug Size, 12mm OD Sidearms
SDP
Supplier:  Chemglass Life Sciences CG44605
Encompass_Preferred
Stopcock, Double Oblique Bore, PTFE Plug, 6mm Bore, 20/65 Plug Size, 12mm OD Sidearms. A 1:5 taper PTFE stopcock with two parallel diagonal bores.
Catalog No. CG44605