Learn More
Med Vet International STPLS/SRG-CLS/20/CRT/24 CRTSCT
Supplier:  Med Vet International JOR800X
Encompass_Preferred
Surgi-Close Skin Staplers Wound closure with the most economical external stapler just became easier Over 80 percent of all human surgical wound closures involve the use of skin staplers In veterinary surgery less than 10 percent of surgeries utilize skin staplers Why is there such a wide difference in usage The use of skin staplers has been limited in veterinary surgery due to the high cost of disposable human skin staplers The use of sutures with needle packs has been less expensive to close the skin (average cost ranges 4- 5) The introduction of the Surgi- Close stapling system addresses the specific needs of the veterinary surgeon Benefits Better apposition of skin edges with better cosmetic results Less reactivity of stainless steel staples Significantly faster wound closure Ease of learning proper technique Features 20-pack staple cartridges cost less than a pack of needle suture and is a hugesavings over conventional throw away skin staples
Catalog No. 50-004-95596