Learn More
Mascon Medical Straight Centrifuge Tube, Polypropylene, 50ml, 25EA/Bag, 20Bags/Case
SDP
Supplier:  Mascon Medical FSCFTUBEST50
Encompass_Preferred
Mascon Medical™ Straight Centrifuge Tubes are engineered to provide a high strength product with vast temperature stability, ensuring outstanding performance in your critical applications. These 50ml free-standing, skirted bottom tubes with flat caps are ideal for cell centrifugation, pelleting, separation by density gradients. These leak proof centrifuge tubes are made of ultra-clear polypropylene, have white printed graduations with a large white marking strip for labeling and are Non-Pyrogenic and DNase/RNase free.
Specifications:
• Volume: 50ml
• Qty. / Case: 500
• Material: Polypropylene
• Cap Material: HDPE
• Color: Clear with Blue Cap
• Packaging Format: 25EA/Bag, 20Bags/Case
Features:
• Non-Pyrogenic
• DNase/RNase free
• Straight
• Free-standing, Skirted Bottom
• Flat Cap
• Leak proof
Catalog No. NC3897065