Learn More
Grainger Straight Cut Profile Router Bit 3/8
Supplier:  Grainger 16Y545
Encompass_Preferred
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router accepts 1/4" and 1/2" shank cutters up to 1-1/2" in diameter. It has a soft grip handle and depth stop for making precision cuts. The router comes with a 1/4" collet, chip deflector, wrenches, and carrying case. This versatile tool is ideal for trimming laminates, cutting grooves, slots and decorative edges.
Catalog No. 50-276-6270