Learn More
Grainger Straight Cut Profile Router Bit 1
Supplier:  Grainger 16Y571
Encompass_Preferred
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for cutting straight edges and profiles in wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router comes with a 1/4" and 1/2" collet, allowing bits up to 1/2" shank diameter. It features a micro-adjustable depth stop and a clear Lexan sub-base for visibility. The router has a soft start motor and electronic speed control. It weighs 4.4 lbs.
Catalog No. 50-276-5547