Learn More
Bellco Glass, Inc. Straight Harvesting Tube,10L 1/4" OD - Ra 32・Finish
SDP
Supplier:  Bellco Glass, Inc. 196490030
Encompass_Preferred
Straight Harvesting Tube for Omni Vessels. Material: 316l stainless steal with ra finish of 32 inch or better, inside and out. MADE IN THE USA. Dimensions: od x id:1/4" x 5/32" x 10.0" long
Catalog No. 50-194-4924