Learn More
Fine Science Tools Moria Ultra Fine Forceps, MC40, Straight, Inox, 0.01 x 0.005 mm, 11 cm
SDP
Supplier:  Fine Science Tools 1137040
Encompass
Style: MC40/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.01 x 0.005mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC9100523