Learn More
Fine Science Tools Semken Forceps, Stainless Steel, Straight, 13 cm
SDP
Supplier:  Fine Science Tools 1100813
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.3 x 1mm/ Alloy / Material: Stainless Steel/ Length: 13cm
Catalog No. NC9602961