Learn More
Aat Bioquest StrandBrite Green Fluorimetric RNA Quantitation Kit (100 Tests)
SDP
Supplier:  Aat Bioquest 17656
Encompass_Preferred
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis S1 nuclease assays RNase protection assays cDNA library preparation reverse transcription PCR and differential display PCR. This catalog is for 100 Tests.
Catalog No. 50-224-9778