Learn More
Grainger Strap Eye Fitting Steel 3/32 in dia.
Supplier:  Grainger 16X645
Encompass_Preferred
The LOOS Eye End Fitting is a 3/32 inch compatible eye end fitting made of carbon steel with a zinc finish. It has a maximum pressure rating of 3000 psi and temperature rating of 300 degrees Fahrenheit. This fitting connects to 3/32 inch OD rigid metal tubing and has a hex head for tightening. It can be used for instrumentation, automation, and other low pressure pneumatic applications. The LOOS eye end fitting is RoHS compliant.
Catalog No. 50-276-5680