Learn More
Grainger Strapping Seal Open Seal RglrDuty PK250
Supplier:  Grainger 16P030
Encompass_Preferred
The PAC STRAPPING PRODUCTS PK-250 is a polyester strapping tool kit. It includes a sealer, dispenser, tensioner, and cutter for applying and securing polyester strapping. The sealer creates secure seals up to 5mm wide. The dispenser holds up to 250 feet of strapping. The tensioner provides up to 250 lbs of tension. The cutter cuts straps cleanly. This kit contains the essential tools for strapping and securing items with polyester strapping. It is suitable for warehouses, shipping departments, and other industrial/commercial uses. The components are made from steel and high impact polymer for durability.
Catalog No. 50-276-4082