Learn More
Grainger Strapping Sealer Manual Heavy Duty
Supplier:  Grainger 16P077
Encompass_Preferred
The Strapping Sealer from Strapack is a handheld strapping tool that seals polypropylene strapping up to 3/4" wide. It features a compact ergonomic design with rubber grip handles for comfort and control. The sealer creates a strong seal using heat to melt the strapping. It has adjustable temperature control up to 400°F. The sealer requires 110V power and comes with a 6 foot cord. It weighs 2 lbs. This is an industrial strength strapping sealer ideal for securing and bundling items for shipping and storage.
Catalog No. 50-276-4207