Learn More
Agilent Technologies STRATAPREPPLASMDMINIPREPKIT
Supplier:  Agilent Technologies 400761
Encompass
StrataPrep Plasmid Miniprep Kit provides a rapid, phenol-free method for purifying pure plasmid DNA. It yields up to 20microgram of plasmid DNA from 1.5 to 3.0mL of culture Accommodates all sizes of DNA from 2.9-kb plasmids to 45-kb cosmids
Catalog No. 50-125-657