Learn More
Cayman Chemical STRculIc AcId methyl esTR 10mg
SDP
Supplier:  Cayman Chemical 2673610 MG
Encompass_Preferred
A methyl ester form of sterculic acid; inhibits the growth of, and is toxic to, the bacteria R. opacus; decreases the fatty acid content, increases the ratio of palmitate to other fatty acids, and decreases the levels of stearate and oleate in R. opacus at 0.25 or 0.5 mM; has a synergistic effect on increased tumor growth induced by aflatoxin Q1 in rainbow trout at 50 ppm
Catalog No. 50-293-4398