Learn More
IBA LifeSciences Strep-Tactin HRP conjugate; 0.5 ml
Supplier:  IBA LifeSciences 21502001
Encompass_Preferred
The streptavidin variant Strep-Tactin is available as conjugate with horseradish peroxidase (HRP) for fast and directed detection of N-terminally, C-terminally, or internally tagged Strep-tagII or Twin-Strep-tag fusion proteins transferred to a blotting membrane or captured on a microplate. Both conjugates are suitable for chromogenic reaction, whereas Strep-Tactin HRP is applicable for chemiluminescence reaction as well.Depending on the host organism, protein extracts may contain small amounts of biotinylated proteins, e.g., the biotin carboxyl carrier protein (BCCP, 22 kDa ) of E. coli. Biotin Blocking Buffer is recommended for blocking of such biotinylated proteins on western blots prior to detection of Strep-tagII or Twin-Strep-tag fusion proteins with Strep-Tactin HRP to prevent the detection of false positive bands. For this purpose, Biotin Blocking Buffer contains the tetrameric biotin binding protein avidin, which selectively binds biotinylated proteins.
Catalog No. NC9523094