Learn More
Neuromics, Inc Strep-Tactin Superflow 50% Suspension, 50 ml is an immobilized streptaviding variant especially optimized for purification of proteins.
Supplier:  Neuromics, Inc 21206025
Encompass
Immobilized streptavidin variant called Strep-Tactin (5 mg/ml resin) especially optimized for purification of Strep-tagII and Twin-Strep-tag fusion proteins*.
Catalog No. NC2071876