Learn More
Agilent Technologies Streptavidin-Allophycocyanin, 10 mg
Supplier:  Agilent Technologies PJ25S10
Encompass
Streptavidin-Allophycocyanin, 10 mg (formerly ProZyme). GT5 APC (Spirulina platensis) is crosslinked and conjugated to Streptavidin (Streptomyces avidinii). The conjugate has been purified to remove unreacted components in order to improve sensitivity and reduce nonspecific binding. As a final step, the product was filtered using a 0.45 µm membrane and stored in a sterile container.
Catalog No. NC2882734