Learn More
Enzo Life Sciences Streptavidin (AP Conjugate) (1ml)
SDP
Supplier:  Enzo Life Sciences ENZPRT1191000
Encompass_Preferred
As part of the need to increase the sensitivity of your sandwich ELISA, this streptavidin AP conjugate can be used with your sandwich ELISA that utilizes a biotinylated detector antibody and the AMP'D ELISA Signal Amplification kit (ENZ-KIT-100) which requires the presence of the AP as the conjugate. As part of the need to increase the sensitivity of your sandwich ELISA, this streptavidin AP conjugate can be used with your sandwich ELISA that utilizes a biotinylated detector antibody and the AMP'D ELISA Signal Amplification kit (ENZ-KIT-100) which requires the presence of the AP as the conjugate. Formulation: Liquid (50 μg/ml). In 0.025M TRIS-HCl, pH 8.0, containing 0.075M sodium chloride, 0.05mM ZnSO4, 2.45mM MgCl2, 0.045% sodium azide, 0.05% BSA and 50% Stabilzyme-AP. Source: Native. Purified from Streptomyces avidinii (SA) and calf intestine (AP). Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-201-3913