Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin AP conjugate
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11089161001
Encompass
Streptavidin is covalently coupled to alkaline phosphatase (AP) for the preparation of streptavidin-AP-conjugate.Alkaline PhosphataseAlkaline Phosphatase is often used as a label in enzyme immunoassays. AP converts the sensitive substrate X-phosphate/NBT into a stable water soluble product.
Catalog No. 50-290-5381