Learn More
Abcam Streptavidin Atto 542 Monovalent Antibody Labeling Kit, 50UG
Supplier:  Abcam AB27218850UG
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using Streptavidin Atto 542 Monovalent Antibody Labeling Kit ab272188. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6039