Learn More
Apexbio Technology LLC Streptavidin Beads C3,1ml
SDP
Supplier:  Apexbio Technology LLC K13021
Encompass_Preferred
APExBIO Streptavidin Beads C3 are ideal for nucleic acid applications (specifically for samples with a high chaotropic salt concentration), immunoassays involving small biotinylated antigens and applications that are not compatible with bovine serum albumin (BSA).Add the beads to a sample containing biotinylated molecules, e.g. peptides or oligonucleotides. During a short incubation, the biotinylated molecule binds to the beads. The molecule-bead complex can be separated by using a magnet. Capture, washing, and detection can be optimized for manual or automated use. With indirect target capture, mix the biotinylated molecule with the sample to capture the molecule-target complex before adding the beads. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0034