Learn More
Streptavidin, Immobilized on Agarose CL-4B, For affinity chromatography, MilliporeSigma™ Supelco™
streptavidin attached to 4% beaded cross-linked agarose via a C6-spacer (1.2 mg streptavidin/ml packed gel)
$291.00 - $1087.00
Specifications
Grade Affinity Chromatography
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-4376
View Documents
MilliporeSigma Supelco
858811ML
1 mL
Each for $291.00
Add to Cart
11-100-3365
View Documents
MilliporeSigma Supelco
858815ML
5 mL
Each for $1,087.00
Add to Cart
Description
Description
Isolation of cell surface glycoprotein by biotinylated concanavalin A and immobilized streptavidin.
Specifications
Specifications
Affinity Chromatography