Learn More
Streptavidin, Immobilized on Agarose CL-4B, For affinity chromatography, MilliporeSigma™ Supelco™
streptavidin attached to 4% beaded cross-linked agarose via a C6-spacer (1.2 mg streptavidin/ml packed gel)
Supplier:  MilliporeSigma Supelco 858815ML
 View more versions of this product
Catalog No. 11-100-3365
Add to Cart
Description
Description
Isolation of cell surface glycoprotein by biotinylated concanavalin A and immobilized streptavidin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Affinity Chromatography
5 mL
Product Suggestions
Product Suggestions