Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin Magnetic Part
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11641778001
Encompass
The Streptavidin Magnetic Particles are polydisperse paramagnetic particles that are designed for the fast separation of a variety of biotin-labeled molecules. The particles are supplied in a suspension that contains 10mg particles per ml in 50mM Hepes 0.1% bovine serum albumin (BSA) 0.01% methyl-isothiazolone pH 7.4.Binding capacities per mg streptavidin magnetic particles1800 pmol free biotin150 pmol biotin-labeled oligonucleotide10 pmol biotin-labeled dsDNA fragment
Catalog No. 50-290-5172