Learn More
AKADEUM LIFE SCIENCES INC Akadeum STREPTAVIDIN MICROBUBBLES
SDP
Supplier:  AKADEUM LIFE SCIENCES INC 11110000
Encompass
Akadeum’s Streptavidin Microbubbles are high-buoyancy microspheres coated in streptavidin. These microbubbles leverage the biotin-streptavidin binding complex and can be used for binding biotinylated target molecules like antibodies, proteins, DNA, and more, allowing for highly-specific and customized applications. Akadeum’s Streptavidin Microbubbles are gently mixed into the sample where they bind to the biotinylated targets. Using their inherent buoyancy, the microbubbles quickly and gently float the biotinylated analytes to the surface for collection or removal. No magnet required. Capacity: 12.5 mLs.
Catalog No. NC1400588