Learn More
ATCC Streptococcus agalactiae; Quantitative DNA
Supplier:  ATCC BAA611DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-2665