Learn More
ATCC Streptococcus dysgalactiae; subsp. equisimilis
Supplier:  ATCC 12394
Encompass
Streptococcus dysgalactiae subsp. equisimilis grouping strain D166B [CIP 104339] is a whole-genome sequenced bacterial strain. This culture is in Lancefield's group G and can be used as a control strain for Group G Streptococcus (GGS).
Catalog No. 50-238-3555