Learn More
ATCC Streptococcus dysgalactiae; subsp. equisimilis
Supplier:  ATCC 12388
Encompass
Streptococcus dysgalactiae subsp. equisimilis grouping strain C74 is a whole-genome sequenced strain that was isolated from a throat culture. The culture was deposited by RC Lancefield and has application as a control strain for Group C Streptococcus (GCS).
Catalog No. 50-238-3552
 Restricted Item