Learn More
ATCC Haemophilus influenzae; type b
Supplier:  ATCC 51654
Encompass
Haemophilus influenzae strain ES2593 [Eagan mutant] is a whole-genome sequenced bacterial strain with applications in respiratory disease research. This strain has genotype ES hktE7::mini-Tn10Cm and is typed as serotype b.
Catalog No. 50-238-0234