Learn More
ATCC Streptococcus mutans
Supplier:  ATCC 35668
Encompass
Streptococcus mutans strain LRA 28 02 81 has applications in water testing and as a quality control strain for API products.
Catalog No. 50-238-3159