Learn More
TOKU-E Pristinamycin, 270076-60-3, 50 mg
Supplier:  TOKU-E P01850MG
Encompass_Preferred
streptogramin antibiotic, C45H54N8O10 (PI)/C28H35N3O7 (PII) Useful in microbiology and cell culture. Mixture of Pristinamycin 1A and IIA.
Catalog No. 50-255-1763